Alfa Aesar™ Zirconium wire, 0.25mm (0.01 in.) dia., 99.2% (metals basis excluding Hf), Hf 4.5% max

Manufacturer: Alfa Aesar™ 43334G6

Quantity 10m

 View more versions of this product

Catalog No. AA43334G6

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7440-67-7
    MDL Number MFCD00011303
    Chemical Name or Material Zirconium wire, 0.25mm (0.01 in.) dia.

    Specifications

    CAS 7440-67-7
    Physical Form Wire
    Weight ≈0.32g/m
    TSCA TSCA
    MDL Number MFCD00011303
    Odor Odorless
    Quantity 10m
    Chemical Name or Material Zirconium wire, 0.25mm (0.01 in.) dia.
    Assay 99.2%, (metals basis excluding Hf), Hf 4.5% max

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit