Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zirconyl chloride hydrate, 99.99%, (trace metal basis), ACROS Organics™

Click to view available options
Quantity:
10g,10g
Packaging:
Glass bottle,Glass bottle
15461-27-5
Cl2H4O2Zr
198.154
MFCD00149897
VBTPZBJEHPOUOI-UHFFFAOYSA-N
zirconyl chloride hydrate,zirconium dichloride oxide hydrate,zirconium, dichlorooxo-, monohydrate,zirconium oxychloride hydrate,cl2ozr.h2o,zirconium dichloride oxide hydrate, puratronic,zirconyl chloride hydrate trace metals basis
16211499
oxozirconium;hydrate;dihydrochloride
O.O=[Zr].Cl.Cl
15461-27-5
Cl2H4O2Zr
198.154
MFCD00149897
VBTPZBJEHPOUOI-UHFFFAOYSA-N
zirconyl chloride hydrate,zirconium dichloride oxide hydrate,zirconium, dichlorooxo-, monohydrate,zirconium oxychloride hydrate,cl2ozr.h2o,zirconium dichloride oxide hydrate, puratronic,zirconyl chloride hydrate trace metals basis
16211499
oxozirconium;hydrate;dihydrochloride
O.O=[Zr].Cl.Cl
For ultra trace analysis
Quantity 10g,10g
Packaging Glass bottle,Glass bottle
Assay Percent Range 99.99%
Linear Formula Zr(Cl)2O·xH2O
Formula Weight 178.13
Grade Trace Metal Basis
Chemical Name or Material Zirconyl chloride hydrate

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger