Learn More
Coast Zithion-X AA Rechargeable Batteries
Click to view available options
Quantity:
12/Pk.
4/Pk.
8/Pk.
Description
Includes
Pack of 12
- COAST ZITHION-X™ rechargeable lithium batteries change the game in responsible, portable power technology.
- Advanced engineering provides a comprehensive upgrade compared with other sources of battery power by delivering long-lasting run times for a clear performance advantage over standard rechargeable lithium and NiMH batteries.
- To mimic the broad compatibility of alkaline batteries, proprietary COAST ZX Optimized Voltage™ allows ZITHION-X batteries to modulate their power supply up or down based on the need and prevent flickering, overheating, and damage to products.
- The leak-resistant battery casing prevents corrosion during use, matching performance with protection.
- Connect conveniently via USB-C to recharge quickly (in 2.5 hours or less for AA and 1.25 hours or less for AAA) and forget about special charging cables and cradles. Every ZITHION-X can be recharged at least 1000 times, resulting in significant cost-savings compared with single-use disposable batteries while keeping hundreds of batteries out of the landfill.
Specifications
Specifications
| Product Type | Rechargeable Battery |
| Battery Type | AA |
| Material | Lithium-ion |
| Cell Type | Spring Terminal |
| Quantity | 12/Pk. |
| Voltage | 1.5 V |