Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris™ ZK 164015

Potent estrogen receptor antagonist

Manufacturer:  Tocris™ 2183/10

 View more versions of this product

Catalog No. 21-831-0


  Check Availability
Add to cart

Disclaimers:

For Research Use Only


Specifications

Description

Potent estrogen receptor silent antagonist. Inhibits 17β-estradiol stimulation of luciferase activity (IC50 = 0.025µM); potently inhibits the growth of estrogen-sensitive human MCF-7 breast cancer cells in vitro (IC50 ~ 1nM).
  • Chemical Name: 2-(4-Hydroxyphenyl)-3-methyl-1-[10-(pentylsulfonyl)decyl]-1H-indol-5-ol
  • Formula: C30H43NO4S
  • CAS Number: 177583-70-9
  • Solubility: Soluble to 25mM in ethanol and to 100mM in DMSO
  • Purity: >99
Description & Specifications

Specifications

ZK 164015
>99
10mg
ZK 164015
513.73
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit