Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZK 200775, Tocris Bioscience™

Catalog Number p-4784860
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
161605-73-8
C14H15F3N3O6P
409.258
WZMQMKNCWDCCMT-UHFFFAOYSA-N
fanapanel, mpqx, fanapanel inn, unii-e3ap71em0o, e3ap71em0o, 7-morpholin-4-yl-2,3-dioxo-6-trifluoromethyl-4h-quinoxalin-1-yl methylphosphonic acid, dsstox_cid_26093, dsstox_rid_81332, dsstox_gsid_46093, 3,4-dihydro-7-morpholino-2,3-dioxo-6-trifluoromethyl-1 2h-quinoxalinyl methyl phosphonic acid
208953
[7-morpholin-4-yl-2,3-dioxo-6-(trifluoromethyl)-4H-quinoxalin-1-yl]methylphosphonic acid
C1COCCN1C2=CC3=C(C=C2C(F)(F)F)NC(=O)C(=O)N3CP(=O)(O)O
This item is not returnable. View return policy
161605-73-8
C14H15F3N3O6P
409.258
WZMQMKNCWDCCMT-UHFFFAOYSA-N
fanapanel, mpqx, fanapanel inn, unii-e3ap71em0o, e3ap71em0o, 7-morpholin-4-yl-2,3-dioxo-6-trifluoromethyl-4h-quinoxalin-1-yl methylphosphonic acid, dsstox_cid_26093, dsstox_rid_81332, dsstox_gsid_46093, 3,4-dihydro-7-morpholino-2,3-dioxo-6-trifluoromethyl-1 2h-quinoxalinyl methyl phosphonic acid
208953
[7-morpholin-4-yl-2,3-dioxo-6-(trifluoromethyl)-4H-quinoxalin-1-yl]methylphosphonic acid
C1COCCN1C2=CC3=C(C=C2C(F)(F)F)NC(=O)C(=O)N3CP(=O)(O)O

Competitive non-NMDA iGluR antagonist

Competitive AMPA/kainate antagonist. In rat cortical membranes, displays high affinity for [3H]-AMPA (Ki = 120 nM) and [3H]-CNQX (Ki = 32 nM) binding sites and low affinity for kainate and NMDA channel-associated binding sites (IC50 values range from 2.5 to 11 μM). Inhibits currents induced by AMPA, kainate and NMDA with IC50 values of 21 nM, 27 nM, and > 1 μM respectively. Displays anxiolytic, anticonvulsant and muscle relaxant activity in vivo.
Quantity 10 mg
Formula Weight 409.25
Percent Purity >99%
Chemical Name or Material ZK 200775
Recommended Storage Desiccate at +4°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit