Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZK 93423 hydrochloride, Tocris Bioscience™

Catalog Number 199450
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
1216574-52-5
C23H23ClN2O4
426.897
KHMDBPAODAAMBQ-UHFFFAOYSA-N
zk 93423 hydrochloride, 4-methoxymethyl-6-phenylmethoxy-9h-pyrido 3,4-b indole-3-carboxylic acid ethyl ester hydrochloride, 4-methoxymethyl-6-phenylmethoxy-9h-pyrido 3,4-b indole-3-carboxylic acid ethyl ester, 9h-pyrido 3,4-b indole-3-carboxylicacid, 4-methoxymethyl-6-phenylmethoxy-, ethyl ester
56972166
ethyl 4-(methoxymethyl)-6-phenylmethoxy-9H-pyrido[3,4-b]indole-3-carboxylate;hydrochloride
CCOC(=O)C1=NC=C2C(=C1COC)C3=C(N2)C=CC(=C3)OCC4=CC=CC=C4.Cl
This item is not returnable. View return policy
1216574-52-5
C23H23ClN2O4
426.897
KHMDBPAODAAMBQ-UHFFFAOYSA-N
zk 93423 hydrochloride, 4-methoxymethyl-6-phenylmethoxy-9h-pyrido 3,4-b indole-3-carboxylic acid ethyl ester hydrochloride, 4-methoxymethyl-6-phenylmethoxy-9h-pyrido 3,4-b indole-3-carboxylic acid ethyl ester, 9h-pyrido 3,4-b indole-3-carboxylicacid, 4-methoxymethyl-6-phenylmethoxy-, ethyl ester
56972166
ethyl 4-(methoxymethyl)-6-phenylmethoxy-9H-pyrido[3,4-b]indole-3-carboxylate;hydrochloride
CCOC(=O)C1=NC=C2C(=C1COC)C3=C(N2)C=CC(=C3)OCC4=CC=CC=C4.Cl

Potent benzodiazepine agonist

Potent benzodiazepine receptor agonist (IC50 = 1 nM). Non-selective between α1-, α2-, α3- and α5-subunit containing GABAA receptors (Ki values are 4.1, 4.2, 6 and 4.5 nM for inhibition of [3H]Ro15-1788 binding to human recombinant α1β3γ2, α2β3γ2, α3β3γ2 and α5β3γ2 receptors respectively). Anxiolytic following systemic administration in vivo.
Quantity 50 mg
Formula Weight 426.9
Percent Purity >98%
Chemical Name or Material ZK 93423 hydrochloride
Recommended Storage Desiccate at Room Temperature
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit