Learn More
ZK 93426 hydrochloride, Tocris
Potent, competitive benzodiazepine antagonist
Manufacturer:  R&D Systems 1996/50
Quantity 50mg
 View more versions of this product
Catalog No. 19-965-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>99%
348.82
ZK 93426 hydrochloride
C18H20N2O3·HCl
4-Methyl-5-(1-methylethoxy)-9H-pyrido[3, 4-b]indole-3-carboxylic acid ethyl ester hydrochloride
Specifications
1216792-30-1
C18H20N2O3·HCl
4-Methyl-5-(1-methylethoxy)-9H-pyrido[3, 4-b]indole-3-carboxylic acid ethyl ester hydrochloride
ZK 93426 hydrochloride
>99%
348.82
50mg
Safety and Handling
Recommended Storage : Desiccate at Room Temperature