Learn More
Tocris Bioscience™ ZM 226600 Available on GSA/VA Contract for Federal Government customers only.
Kir6 (KATP) channel opener
Manufacturer:  Tocris Bioscience™ 0882/10
 View more versions of this product
Catalog No. 08-821-0
Add to cart
Description
Description
Potent Kir6 (KATP) channel opener (EC50 = 0.5µM), devoid of antiandrogen properties.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
N-(4-Phenylsulfonylphenyl)-3,3,3-trifluoro-2-hydroxy-2-methylpropanamide
C16H14F3NO4S
10mg
147695-92-9
Observed MW: 373.35
Soluble to 100mM in DMSO
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT