Learn More
Tocris Bioscience™ ZM 226600
Kir6 (KATP) channel opener
Manufacturer:  Tocris Bioscience™ 0882/50
 View more versions of this product
Catalog No. 08-825-0
Add to cart
Description
Description
Potent Kir6 (KATP) channel opener (EC50 = 0.5µM), devoid of antiandrogen properties.
Specifications
Specifications
N-(4-Phenylsulfonylphenyl)-3,3,3-trifluoro-2-hydroxy-2-methylpropanamide
C16H14F3NO4S
50mg
147695-92-9
Observed MW: 373.35
Soluble to 100mM in DMSO
Safety and Handling
Safety and Handling
Recommended Storage : Store at RT