Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ ZM 306416 hydrochloride Available on GSA/VA Contract for Federal Government customers only.

Inhibitor of VEGF receptor tyrosine kinase

$101.50 - $218.50

Specifications

Assay Percent Range >99%
Formula Weight Observed MW: 370.21
Chemical Name or Material 4-[(4'-Chloro-2'-fluoro)phenylamino]-6,7-dimethoxyquinazoline hydrochloride
Molecular Formula C16H13N3O2FCl.HCl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
24-991-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
2499/10
10mg Each for $218.50
Add to cart
 
24-991-R Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
2499/1
1mg Each for $101.50
Add to cart
 
Description

Description

Inhibitor of vascular endothelial growth factor (VEGF) receptor tyrosine kinase. Inhibits activity of KDR and FLT VEGF receptors (IC50 values are 100 nM and 2 µM respectively) and displays > 3-fold selectivity over FGFR-1. Orally active in vivo.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

>99%
Observed MW: 370.21
4-[(4'-Chloro-2'-fluoro)phenylamino]-6,7-dimethoxyquinazoline hydrochloride
C16H13N3O2FCl.HCl
Safety and Handling

Safety and Handling

Recommended Storage : Desiccate at RT

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit