Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ ZM-306416 hydrochloride, 98%

Click to view available options
Quantity:
1mg
10mg
690206-97-4
C16H13ClFN3O2
333.747
MFCD08703133
YHUIUSRCUKUUQA-UHFFFAOYSA-N
n-4-chloro-2-fluorophenyl-6,7-dimethoxyquinazolin-4-amine,anilinoquinazoline deriv. 4,d0c6ki,n-2-fluoro-4-chlorophenyl-6,7-dimethoxyquinazoline-4-amine,n-4-chloro-2-fluorophenyl-6,7-dimethoxy-4-quinazolinamine
5329006
N-(4-chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine
COC1=C(C=C2C(=C1)C(=NC=N2)NC3=C(C=C(C=C3)Cl)F)OC
690206-97-4
C16H13ClFN3O2
333.747
MFCD08703133
YHUIUSRCUKUUQA-UHFFFAOYSA-N
n-4-chloro-2-fluorophenyl-6,7-dimethoxyquinazolin-4-amine,anilinoquinazoline deriv. 4,d0c6ki,n-2-fluoro-4-chlorophenyl-6,7-dimethoxyquinazoline-4-amine,n-4-chloro-2-fluorophenyl-6,7-dimethoxy-4-quinazolinamine
5329006
N-(4-chloro-2-fluorophenyl)-6,7-dimethoxyquinazolin-4-amine
COC1=C(C=C2C(=C1)C(=NC=N2)NC3=C(C=C(C=C3)Cl)F)OC
Quantity 10mg
Color White
Formula Weight 370.21
Physical Form Solid
Percent Purity 98%
Chemical Name or Material ZM-306416 hydrochloride