Learn More
Tocris Bioscience™ ZM 306416 hydrochloride, Tocris Available on GSA/VA Contract for Federal Government customers only.
Inhibitor of VEGF receptor tyrosine kinase
Manufacturer:  Tocris Bioscience™ 2499/10
 View more versions of this product
Catalog No. 24-991-0
Add to cart
This item is not returnable. View return policy
Description
Description
Inhibitor of vascular endothelial growth factor (VEGF) receptor tyrosine kinase. Inhibits activity of KDR and FLT VEGF receptors (IC 50 values are 100 nM and 2 µM respectively) and displays > 3-fold selectivity over FGFR-1. Orally active in vivo.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
4-[(4'-Chloro-2'-fluoro)phenylamino]-6,7-dimethoxyquinazoline hydrochloride
>99%
Observed MW: 370.21
Soluble to 10mM in DMSO
196603-47-1
C16H13N3O2FCl.HCl
10mg
Safety and Handling
Safety and Handling
Recommended Storage : Desiccate at RT