Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZMAT1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549301

Catalog No. PA549301

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a protein containing Cys2-His2 (C2H2)-type zinc fingers, which are similar to those found in the nuclear matrix protein matrin 3. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq, Jan 2012]
Specifications

Specifications

ZMAT1
Polyclonal
PBS with 0.09% sodium azide
Q5H9K5
Zinc finger matrin-type protein 1, ZMAT1, KIAA1789
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84460
Western Blot
Unconjugated
ZMAT1
Liquid
ZMAT1
KLH conjugated synthetic peptide between 548-577 amino acids from the C-terminal region of human ZMAT1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit