Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZMIZ2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA531271
Description
Recommended positive controls: IMR32. Predicted reactivity: Mouse (92%), Rat (93%), Bovine (92%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ZIMP7, also known as ZMIZ2, is a novel PIAS (protein inhibitor of activated signal transducer and activator of transcription)-like protein and a transcriptional coactivator. ZIMP7 is expressed most abundantly in testis. The C-terminal proline-rich domain possesses a significant intrinsic transcriptional activity and this activity is inhibited by the N-terminus in the full-length ZIMP7. ZIMP7 and the related protein ZIMP10 interact with PIAS3 and enhances Androgen Receptor (AR)- mediated transcription. The interaction between ZIMP7 and SWI/SNF complex suggests a possible role for ZIMP7 in chromatin modification.
Specifications
ZMIZ2 | |
Polyclonal | |
Unconjugated | |
ZMIZ2 | |
D11Bwg0280e; HRIHFB2007; hZIMP7; KIAA1886; NET27; PIAS-like protein Zimp7; TRAFIP20; Zimp7; zinc finger MIZ domain-containing protein 2; zinc finger MIZ-type containing 2; zinc finger, MIZ-type containing 2; Zmiz2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
83637 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.93 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
Q8NF64 | |
ZMIZ2 | |
Recombinant fragment corresponding to a region within amino acids 569 and 818 of Human ZMIZ2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction