Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZMYM6 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP326106PCP
Description
ZMYM6 Polyclonal antibody specifically detects ZMYM6 in Human samples. It is validated for Western BlotSpecifications
ZMYM6 | |
Polyclonal | |
Western Blot | |
Buster2, KIAA1353, MYM, MYM-type containing 6, zinc finger MYM-type protein 6, zinc finger, MYM-type 6 | |
Produced in rabbits immunized with E. coli-derived Human ZMYM6 fragment | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
PerCP | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
9204 | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction