Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

KRBOX4 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548762

Catalog No. PA548762

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This encodes a zinc finger protein with an N-terminal KRAB (Kruppel-associated) domain found in transcriptional repressors. This gene is located in a region of the X chromosome thought to be involved in nonsyndromic X-linked mental retardation. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Aug 2011]
Specifications

Specifications

KRBOX4
Polyclonal
PBS with 0.09% sodium azide
Q5JUW0
KRAB domain-containing protein 4, KRAB box domain-containing protein 4, KRBOX4, ZNF673
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
55634
Western Blot
Unconjugated
KRBOX4
Liquid
KRBOX4
KLH conjugated synthetic peptide between 96-125 amino acids from the C-terminal region of human ZN673.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit