Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNDR1 Mouse anti-Human, DyLight 550, Clone: OTI1G2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP274930R
Description
ZNDR1 Monoclonal antibody specifically detects ZNDR1 in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.Specifications
ZNDR1 | |
OTI1G2 | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Affinity Purified, Conjugated | |
ZNRD1 | |
Full length recombinant protein of human ZNDR1 (NP_740753) produced in HEK293T cells. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Immunocytochemistry, Immunofluorescence, Western Blot | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
DNA-directed RNA polymerase I subunit RPA12, HTEX-6, hZR14, MGC13376, RNA polymerase I small specific subunit Rpa12, Rpa12, tctex-6, TEX6, transcription-associated zinc ribbon protein, zinc ribbon domain containing 1, zinc ribbon domain containing, 1, Zinc ribbon domain-containing protein 1 | |
Mouse | |
IgG1 | |
0.1 mL | |
Primary | |
30834.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title