Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF100 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18018320UL

 View more versions of this product

Catalog No. NBP18018320


Add to cart

Description

Description

ZNF100 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZNF100
LYOPH
Western Blot 1:1000
NP_775802
FLJ44587, zinc finger protein 100, zinc finger protein 100 (Y1)
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
163227
Human
Western Blot
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
ZNF100
Synthetic peptide directed towards the N terminal of human ZNF100. Peptide sequence MDDPRYGMCPLKGASGCPGAERSLLVQSYFEKGPLTFRDVAIEFSLEEWQ.
63 kDa
20 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge prior to opening. Reconstitute with sterilized PBS to a final concentration of 1mg/ml. Vortex followed by Centrifuge again to pellet the solution.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit