Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF12 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18027620UL

 View more versions of this product

Catalog No. NBP18027620


Add to cart

Description

Description

ZNF12 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZNF12
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
ZNF12
Synthetic peptide directed towards the N terminal of human ZNF12. Peptide sequence: ADECSGCGKSLLHIKLEKTHPGDQAYEFNQNGEPYTLNEESLYQKIRILE
Protein A purified
RUO
Primary
7559
Western Blot
Western Blot 1:1000
NP_057349
GIOT3, Gonadotropin-inducible ovary transcription repressor 3, HZF11, zinc finger protein 11, zinc finger protein 12, Zinc finger protein 325GIOT-3KOX3gonadotropin inducible transcription repressor 3, Zinc finger protein KOX3, ZNF325
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit