Learn More
Abnova™ ZNF131 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00007690T03
Catalog No. 89-967-975
Description
Description
Specifications
Specifications
Western Blot
ZNF131 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 57.9kDa
Transient overexpression cell lysate was tested with anti-ZNF131 antibody (Mfr. No. H00007690-B01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
BC035875.2
pCMV-ZNF131 full-length
100 μL
Human