Learn More
Aviva Systems Biology Corp ZNF134 Antibody - middle region (ARP58103_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58103_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF134. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF134. Alternate names: MGC138499, MGC141970, pHZ-15ApplicationWB. Species reactivity: Human, Pig
Catalog No. 50-014-62238