Learn More
Aviva Systems Biology Corp ZNF138 Antibody - middle region (ARP47604_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47604_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF138. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF138. Alternate names: pHZ-32ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat, Zebrafish
Catalog No. 50-014-59606