Learn More
Aviva Systems Biology Corp ZNF148 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP34606_P050
This is a rabbit polyclonal antibody against ZNF148. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF148. Alternate names: BERF-1, BFCOL1, HT-BETA, ZBP-89, ZFP148, pHZ-52ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-55477