Learn More
ZNF154 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 221611AP
Catalog No. 50-173-3750
Add to cart
Description
Description
ZNF154 may be involved in transcriptional regulation.
Specifications
Specifications
ZNF154
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q13106
KIAA2003, pHZ 92, zinc finger protein 154, ZNF154
Rabbit
IgG
150 μL
-20°C
Primary
7710
Western Blot
Unconjugated
ZNF154
Liquid
ZNF154
ZNF154 Fusion Protein Ag17812
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human