Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF165 Mouse anti-Human, DyLight™ 405, Clone: OTI4C1, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274932V

Catalog No. NBP274932V


Add to cart

Description

Description

ZNF165 Monoclonal antibody specifically detects ZNF165 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF165
OTI4C1
Western Blot
Affinity Purified, Conjugated
ZNF165
Recombinant protein fragment corresponding to amino acids 165-485 of human ZNF165 (NP_003438) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
Cancer/testis antigen 53, CT53Zinc finger and SCAN domain-containing protein 7, zinc finger protein 165, ZSCAN7LD65ZPF165
Mouse
IgG1
0.1 mL
Primary
7718.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit