Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF169 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA559718
Description
Immunogen sequence: KGESGETEGP DSSLRKRPSR ISRTFFSPHQ GDPVEWVEGN REGGTDLRLA QRMSLGGSDT MLKGADTSES GAVIRGNYRL GLSKKSSLFS HQKH Highest antigen sequence identity to the following orthologs: Mouse - 45%, Rat - 45%.
ZNF169 (Zinc finger protein 169) is a 603 amino acid nuclear protein that contains one KRAB domain and thirteen C2H2-type zinc fingers. ZNF169 is highly expressed in kidney and weakly expressed in spleen, liver, small intestine and heart, where it functions as a transcription regulator. The gene encoding ZNF169 maps to a region of human chromosome 9q22, which has been associated with many human diseases such as colon cancer, migraine auras, basal cell carcinoma, Gorlin syndrome and Extraskeletal myxoid chondrosarcoma.
Specifications
ZNF169 | |
Polyclonal | |
Unconjugated | |
ZNF169 | |
1700025J14Rik; 4930429A13Rik; BE688704; RP11-307E17.10-001; Zfp169; Zinc finger protein 169; ZNF169 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
169841 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14929 | |
ZNF169 | |
Recombinant protein corresponding to Human ZNF169. Recombinant protein control fragment (Product #RP-99920). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction