Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF17 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF17 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180203
|
Novus Biologicals
NBP180203 |
100 μL |
Each of 1 for $436.00
|
|
Description
ZNF17 Polyclonal specifically detects ZNF17 in Human samples. It is validated for Western Blot.Specifications
ZNF17 | |
Polyclonal | |
Rabbit | |
Human | |
FLJ40864, FLJ46058, FLJ46615, HPF3, KIAA1947, KOX10, zinc finger protein 17, zinc finger protein 17 (HPF3, KOX 10), zinc finger protein HPF3, zinc finger protein KOX10 | |
ZNF17 | |
IgG | |
Affinity Purified | |
53 kDa |
Western Blot | |
Unconjugated | |
RUO | |
NP_008890 | |
7565 | |
Synthetic peptide directed towards the N terminal of human ZNF17. Peptide sequence NLTCMQGGKDFTGDSDLQQQALHSGWKPHRDTHGVEAFQSGQNNYSCTQC. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title