Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF174 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA124365
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA124365 100 μg
1 options

Catalog No. PIPA124365

Supplier: Thermo Scientific PA124365

Rabbit Polyclonal Antibody

PA1-24365 detects ZNF174 in human samples. PA1-24365 has been successfully used in Western blot procedures. PA1-24365 immunogen corresponds to Polyclonal antibody from rabbits immunized with a synthetic peptide region of human ZNF174. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF174
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Gene ZNF174
Gene Accession No. Q15697
Gene Alias ZSCAN8
Gene Symbols ZNF174
Host Species Rabbit
Immunogen Synthetic peptide region of human ZNF174.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7727
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.