Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF175 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549338

Catalog No. PA549338

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

ZNF175 expression in brain mononuclear phagocytes is a signature for advanced HIV-1 encephalitis. As a transcriptional suppressor, this protein plays a role in macrophage control of viral replication during advanced HIV-1 infection.
Specifications

Specifications

ZNF175
Polyclonal
PBS with 0.09% sodium azide
Q9Y473
Zinc finger protein 175, Zinc finger protein OTK18, ZNF175
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7728
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
ZNF175
Liquid
ZNF175
KLH conjugated synthetic peptide between 150-187 amino acids from the N-terminal region of human ZNF175.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit