Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF177 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ZNF177
Applications Immunofluorescence, Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093504
View Documents
Novus Biologicals
NBP305074100UL
100 μg Each for $320.71
Add to cart
 
NB093505
View Documents
Novus Biologicals
NBP30507420UL
20 μg Each for $138.37
Add to cart
 
Description

Description

ZNF177 Polyclonal antibody specifically detects ZNF177 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence
Specifications

Specifications

ZNF177
Unconjugated
Rabbit
Human, Mouse, Rat
PIGX, zinc finger protein 177
Recombinant fusion protein containing a sequence corresponding to amino acids 50-110 of human ZNF177 (NP_003442.2).
Polyclonal
Affinity purified
Immunofluorescence, Immunohistochemistry
Purified
RUO
PBS with 50% glycerol, pH7.3.
7730
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit