Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF182 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110598
Description
Immunogen sequence: QVDEQIERQH QDDQDKCLLM QVGFSDKKTI ITKSARDCHE FGNILHLSTN LVASIQRPDK HESFGNNMVD NLDLFSRSSA ENKYDNGCAK LFFHTEYEKT NPGMKPYGYK E.
Zinc-finger proteins bind nucleic acids and play important roles in various cellular functions, including cell proliferation, differentiation, and apoptosis. This gene encodes a zinc finger protein, and belongs to the krueppel C2H2-type zinc-finger protein family. It may be involved in transcriptional regulation. Multiple alternatively spliced transcript variants encoding different isoforms have been identified.
Specifications
ZNF182 | |
Polyclonal | |
Unconjugated | |
ZNF182 | |
6330583I20Rik; HHZ150; KOX14; MGC137263 protein; Zfp182; Zinc finger protein 182; zinc finger protein 21; zinc finger protein 21 (KOX 14); zinc finger protein KOX14; ZNF182; ZNF21 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
100911831, 319535, 7569 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.45 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
P17025, Q6P560 | |
Zfp182, ZNF182 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 100-210 of human ZNF182 (NP_001171570.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction