Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

ZNF185 Rabbit anti-Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Specifications

Antigen ZNF185
Dilution Western Blot 1.0 ug/ml
Applications Western Blot
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB125540
View Documents
Novus Biologicals
NBP31031925UL
25 μg
Each for $106.19
N/A
NB125539
View Documents
Novus Biologicals
NBP310319100UL
100 μg
Each for $352.89
N/A
Description

Description

ZNF185 Polyclonal antibody specifically detects ZNF185 in Mouse samples. It is validated for Western Blot
Specifications

Specifications

ZNF185
Western Blot
Purified
RUO
PBS buffer, 2% sucrose
7739
Polyclonal
Affinity purified
Western Blot 1.0 ug/ml
Unconjugated
Rabbit
Mouse
zinc finger protein 185 (LIM domain)
The immunogen is a synthetic peptide directed towards the middle region of MOUSE Znf185. Peptide sequence REHSYVLSAAKKTTSSPTQELQSPFLAKRVDVVDEDVLPEKNQEPPALAR
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit