Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF205 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18030220UL

 View more versions of this product

Catalog No. NBP18030220


Add to cart

Description

Description

ZNF205 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZNF205
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
ZNF205
Synthetic peptide directed towards the N terminal of human ZNF205. Peptide sequence PSKLGEAVPSGDTQESLHIKMEPEEPHSEGASQEDGAQGAWGWAPLSHGS.
Protein A purified
RUO
Primary
7755
Western Blot
Western Blot 1:1000
NP_001026856
zinc finger protein 205, Zinc finger protein 210, ZNF210Zfp13
Rabbit
IgG
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit