Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF217 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178189
Description
ZNF217 Polyclonal specifically detects ZNF217 in Human samples. It is validated for Western Blot, Immunoprecipitation, Knockdown Validated.Specifications
ZNF217 | |
Polyclonal | |
Unconjugated | |
TBS and 0.1% BSA with 0.09% Sodium Azide | |
DKFZp781O2147, KIAA0335, MGC125707, MGC125710, zinc finger protein 518, zinc finger protein 518A, ZNF518 | |
Rabbit | |
Affinity Purified | |
RUO | |
7764 | |
Store at 4C. Do not freeze. |
Western Blot, Immunoprecipitation | |
0.2 mg/mL | |
Western Blot 1:2000-1:10000, Immunoprecipitation 2-10 ug/mg lysate, KnockDown Validated | |
NP_006517.1 | |
ZNF217 | |
The immunogen for this product maps to a region between residue 250 and 300 of human Zinc Finger Protein 217 using the numbering given in entry NP_006517.1 (GeneID 7764). | |
0.1 mL | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction