Learn More
ZNF22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 170631AP
Catalog No. 50-173-1359
Add to cart
Description
Description
ZNF22 encodes a protein that is expressed in the epithelial component of the developing tooth organ during early bud and cap stages as well as in osteoblasts of craniofacial bone and the developing tongue.
Specifications
Specifications
ZNF22
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P17026, Q9ERU2, Q9ERU3
HKR T1, KOX15, KROX26, Zfp422, Zinc finger protein 22, Zinc finger protein KOX15, Zinc finger protein Krox 26, ZNF22, ZNF422
Rabbit
IgG
150 μL
-20°C
Primary
360389, 67255, 7570
Western Blot
Unconjugated
ZNF22
Liquid
Znf22
ZNF22 Fusion Protein Ag10833
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat