Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF227 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP18019420UL

 View more versions of this product

Catalog No. NBP18019420


Add to cart

Description

Description

ZNF227 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZNF227
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Purified
ZNF227
Synthetic peptide directed towards the middle region of human ZNF227. Peptide sequence ESGDCFNKSSFHSYQSNHTGEKSYRCDSCGKGFSSSTGLIIHYRTHTGEK.
30 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Western Blot
Western Blot 1:1000
AAO45835
KIAA1084, zinc finger protein 507
Rabbit
IgG
Protein A purified
RUO
Primary
7770
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit