Learn More
ZNF23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 213221AP
Catalog No. 50-173-3367
Add to cart
Description
Description
ZNF23 May be involved in transcriptional regulation. May have a role in embryonic development. Belongs to the krueppel C2H2-type zinc-finger protein family.
Specifications
Specifications
ZNF23
0.32 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P17027
KOX16, Zfp612, Zinc finger protein 23, Zinc finger protein 359, Zinc finger protein KOX16, ZNF23, ZNF359, ZNF612
Rabbit
IgG
150 μL
-20°C
Primary
234725, 307839, 7571
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot
Unconjugated
ZNF23
Liquid
Zfp612, ZNF23
ZNF23 Fusion Protein Ag14755
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat