Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF230 Mouse anti-Human, Clone: OTI1E2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274934

Catalog No. NBP274934


Add to cart

Description

Description

ZNF230 Monoclonal antibody specifically detects ZNF230 in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

ZNF230
OTI1E2
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
FLJ12515, FLJ14855, zinc finger protein 502
Mouse
IgG1
Immunogen affinity purified
Zinc Finger
Primary
7773
Human
Immunohistochemistry
LYOPH
Immunohistochemistry 1:150
Affinity Purified, Lyophilized
ZNF230
Full length human recombinant protein of human ZNF230 (NP_006291) produced in E.coli.
54.4 kDa
100 μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit