Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF232 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA565027
Description
Immunogen sequence: LEDLEKGLEPE PQVPGPAHGP AQEEPWEKKE SLGAAQEALS IQLQPKETQP FPKSVVTEDG PEPKDKGSLP QPPITEVESQ VFSEKLATDT STFEATSEGT LELQQRNPKA E Highest antigen sequence identity to the following orthologs - mouse 29%, rat 30%.
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF232, also known as Zinc finger and SCAN domain-containing protein 11, is a 417 amino acid protein belonging to the Kruppel C2H2-type zinc-finger protein family. Localized to the nucleus, ZNF232 contains one SCAN box domain and five C2H2-type zinc fingers. Due to the presence of these domains, ZNF232 may be involved in transcriptional regulation. Ubiquitously expressed, ZNF232 is present at high levels in testis, liver and ovary. ZNF232 exists as two isoforms produced by alternative splicing.
Specifications
ZNF232 | |
Polyclonal | |
Unconjugated | |
ZNF232 | |
Zinc finger and SCAN domain-containing protein 11; zinc finger protein 232; ZNF232; ZSCAN11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7775 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UNY5 | |
ZNF232 | |
Recombinant Human ZNF232. Recombinant protein control fragment (Product #RP-105793). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction