Learn More
ZNF232 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 214671AP
Catalog No. 50-173-3435
Add to cart
Description
Description
Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Kruppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. ZNF232, also known as Zinc finger and SCAN domain-containing protein 11, is a 417 amino acid protein belonging to the Kruppel C2H2-type zinc-finger protein family. Localized to the nucleus, ZNF232 contains one SCAN box domain and five C2H2-type zinc fingers. Due to the presence of these domains, ZNF232 may be involved in transcriptional regulation. Ubiquitously expressed, ZNF232 is present at high levels in testis, liver and ovary. ZNF232 exists as two isoforms produced by alternative splicing.
Specifications
Specifications
ZNF232
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9UNY5
zinc finger protein 232, ZNF232, ZSCAN11
Rabbit
IgG
150 μL
-20°C
Primary
7775
Western Blot
Unconjugated
ZNF232
Liquid
ZNF232
ZNF232 Fusion Protein Ag15105
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse