Learn More
Abnova™ ZNF25 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00219749T01
Catalog No. 89-968-108
Add to cart
Description
Description
Specifications
Specifications
Western Blot
ZNF25 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 50.27kDa
Transient overexpression cell lysate was tested with anti-ZNF25 antibody (Mfr. No. H00219749-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
293T
NM_145011.2
pCMV-ZNF25 full-length
100 μL
Human