Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF263 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548786

Catalog No. PA548786


Add to cart

Description

Description

Might play an important role in basic cellular processes as a transcriptional repressor.
Specifications

Specifications

ZNF263
Polyclonal
PBS with 0.09% sodium azide
O14978
Zinc finger protein 263, Zinc finger protein FPM315, Zinc finger protein with KRAB and SCAN domains 12, ZNF263, FPM315, ZKSCAN12
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
10127
Western Blot
Unconjugated
ZNF263
Liquid
ZNF263
KLH conjugated synthetic peptide between 318-346 amino acids from the Central region of human ZNF263.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit