Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF263 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 213261AP150UL
Description
Might play an important role in basic cellular processes as a transcriptional repressor.Specifications
ZNF263 | |
Polyclonal | |
Unconjugated | |
ZNF263 | |
FPM315, zinc finger protein 263, Zinc finger protein FPM315, ZKSCAN12, ZNF263 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10127 | |
-20°C | |
Liquid |
Immunoprecipitation | |
0.25 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14978 | |
ZNF263 | |
ZNF263 Fusion Protein Ag15158 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction