Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF264 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568964
Description
This target displays homology in the following species: Cow: 86%; Dog: 79%; Horse: 86%; Human: 100%; Rabbit: 79%; Rat: 79%.
ZNF264 gene encodes a zinc finger protein and belongs to the krueppel C2H2-type zinc-finger protein family. Zinc finger proteins are often localized in the nucleus, bind nucleic acids, and regulate transcription.
Specifications
ZNF264 | |
Polyclonal | |
Unconjugated | |
ZNF264 | |
KIAA0412; Zinc finger protein 264; ZNF264 | |
Rabbit | |
Affinity chromatography | |
RUO | |
9422 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot, Immunocytochemistry | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O43296 | |
ZNF264 | |
synthetic peptide directed towards the C terminal region of human ZNF264. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction