Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF264 Mouse anti-Human, Clone: OTI8F4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527309

Catalog No. PIMA527309


Add to cart

Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF264
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
O43296
ZNF264; KIAA0412
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9422
Western Blot
OTI8F4
Unconjugated
ZNF264
Liquid
ZNF264
Full length human recombit protein of ZNF264 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit