Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF267 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Supplier: Abnova Corporation PAB30642
Description
Rabbit polyclonal antibody raised against partial recombinant human ZNF267.
- High specificity
- High reactivity
Specifications
ZNF267 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant human ZNF267. | |
In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide). | |
HZF2 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
10308 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunofluorescence, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Immunofluorescence (1-4 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-1:200) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user. | |
Q14586 | |
ZNF267 | |
Recombinant protein corresponding to human ZNF267. | |
100 μL | |
Primary | |
Human | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?
Provide Content Correction