Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF271 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309197100UL
Description
ZNF271 Polyclonal specifically detects ZNF271 in Human samples. It is validated for Western Blot.Specifications
ZNF271 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
HZF7, ZNF271 zinc finger protein 271, ZNFEB | |
The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF271 (NP_006620). Peptide sequence GFILERNPIPVINVAKPSVGFQILLIINEFTLERSLTHVISAIKCLVEDE | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
10778 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction