Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF274 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | ZNF274 |
---|---|
Dilution | Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB125947
|
Novus Biologicals
NBP310523100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
ZNF274 Polyclonal specifically detects ZNF274 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ZNF274 | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human | |
GIOT2, GIOT-2, gonadotropin inducible transcription repressor-2, Gonadotropin-inducible ovary transcription repressor 2, gonadotropin-inducible transcription repressor 2, KOX7DKFZp434F1811, zinc finger protein 44, zinc finger protein 44 (KOX 7), Zinc finger protein 55DKFZp686L21136, Zinc finger protein 58ZNF504, Zinc finger protein KOX7, zinc finger protein ZnFP12, ZNF, ZNF55, ZNF58 | |
The immunogen is a synthetic peptide directed towards the middle region of human ZNF274 (NP_057408). Peptide sequence SHLIRHQRTHTGERPYACNKCGKAFTQSSHLIGHQRTHNRTKRKKKQPTS | |
Affinity purified |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
10782 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title