Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF276 Antibody (PCRP-ZNF276-1A5), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP314066AF350
This item is not returnable.
View return policy
Description
ZNF276 Monoclonal antibody specifically detects ZNF276 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunofluorescenceSpecifications
ZNF276 | |
Monoclonal | |
Alexa Fluor 350 | |
DNA-binding protein, KIAA1710Zfp46, zinc finger protein 436, ZNF | |
Recombinant full-length human ZNF276 protein (Uniprot: Q8N554) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunofluorescence | |
PCRP-ZNF276-1A5 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
92822 | |
Store at 4°C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction