Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF280A Mouse anti-Human, Clone: OTI2H3, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526388

Catalog No. PIMA526388


Add to cart

Description

Description

Zinc-finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc-finger proteins contain a Krueppel-type DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. SUHW1 (Suppressor of hairy wing homolog 1), also known as ZNF280A (Zinc finger protein 280A), 3 ft.OY11.1, ZNF636 or ZNF280, is a 542 amino acid protein that contains four C2H2-type zinc fingers. Localized to the nucleus, SUHW1 is thought to function as a transcription factor that may mediate transcriptional regulation events.
Specifications

Specifications

ZNF280A
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
P59817
Zinc finger protein 280A, 3′OY11.1, Suppressor of hairy wing homolog 1, Zinc finger protein 636, ZNF280A, SUHW1, ZNF280, ZNF636
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
129025
Flow Cytometry, Western Blot
OTI2H3
Unconjugated
ZNF280A
Liquid
ZNF280A
Human recombit protein fragment corresponding to amino acids 1-333 of ZNF280A produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit